missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GET4 Protein
A cDNA sequence encoding the GET4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | GET4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978908
|
enQuireBio™
QP11953-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968908
|
enQuireBio™
QP11953-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15958908
|
enQuireBio™
QP11953-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
GET4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GET4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAAAAM AEQESARNGG RNRGGVQRVE GKLRASVEKG DYYEAHQMYR TLFFRYMSQS KHTEARELMY SGALLFFSHG QQNSAADLSM LVLESLEKAE VEVADELLEN LAKVFSLMDP NSPERVTFVS RALKWSSGGS GKLGHPRLHQ LLALTLWKEQ NYCESRYHFL HSADGEGCAN MLVEYSTSRG FRSEVDMFVA QAVLQFLCLK NKSSASVVFT TYTQKHPSIE DGPPFVEPLL NFIWFLLLAV DGGKLTVFTV LCEQYQPSLR RDPMYNEYLD RIGQLFFGVP PKQTSSYGGL LGNLLTSLMG SSEQEDGEES PSDGSPIELD | |
Greater than 90% as determined by SDS-PAGE. |