missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Growth Hormone 2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 7668-GH-025/CF

247.94 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18369687

  • £320.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Growth Hormone 2 Protein is derived from E. coli. The Recombinant Human Growth Hormone 2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl.
22.4 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
2689
Growth Hormone 2
E. coli-derived human Growth Hormone 2 protein Phe27-Phe217,with an N-terminal Met
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers