missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Growth Hormone 2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£309.93
Specifications
Gene ID (Entrez) | 2689 |
---|---|
Molecular Weight (g/mol) | 22.4 kDa |
Name | Growth Hormone 2 |
Quantity | 25μg |
Source | E. coli-derived human Growth Hormone 2 protein Phe27-Phe217,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18168630
|
R&D Systems™
7668-GH-025 |
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
£309.93
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18369687
|
R&D Systems™
7668-GH-025/CF |
Lyophilized from a 0.2μm filtered solution in HCl. |
£309.93
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human Growth Hormone 2 Protein is derived from E. coli. The Recombinant Human Growth Hormone 2 Protein has been validated for the following applications: Bioactivity.Specifications
2689 | |
Growth Hormone 2 | |
E. coli-derived human Growth Hormone 2 protein Phe27-Phe217,with an N-terminal Met | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
22.4 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |