missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-6 Protein
A New rh IL-6 is Available! The new protein has ∼2 fold better activity and is HEK293 expressed!
Brand: R&D Systems™ 206-IL-001MG/CF
2783.51 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.2-0.8 ng/mL.Specifications
Q75MH2 | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl. | |
M.W. theroretical: 20.9 kDa | |
1 mg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
Unconjugated | |
Reconstitute at 100-200 μg/mL in sterile PBS. | |
IL-6 |
Bioactivity | |
3569 | |
IL-6 | |
E. coli-derived human IL-6 protein Pro29-Met212 | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
IL6 | |
Recombinant | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |