missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-6 Protein

A New rh IL-6 is Available! The new protein has ∼2 fold better activity and is HEK293 expressed!

£386.00 - £3535.00

Specifications

Accession Number Q75MH2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 3569
Molecular Weight (g/mol) M.W. theroretical: 20.9 kDa
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18776806
View Documents
R&D Systems™
206-IL-025/CF
25 μg
£386.00
25µg
Please to purchase this item. Need a web account? Register with us today!
18354837
View Documents
R&D Systems™
206-IL-001MG/CF
1 mg
£3535.00
1mg
Please to purchase this item. Need a web account? Register with us today!
18128360
View Documents
R&D Systems™
206-IL-050/CF
50 μg
£780.00
50µg
Please to purchase this item. Need a web account? Register with us today!
18748906
View Documents
R&D Systems™
206-IL-200/CF
200 μg
£1065.00
200µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.2-0.8 ng/mL.
Specifications

Specifications

Q75MH2
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
M.W. theroretical: 20.9 kDa
E. coli-derived human IL-6 protein Pro29-Met212
<0.10 EU / 1 μg of the protein by the LAL method.
IL6
Recombinant
IL-6
Bioactivity
3569
IL-6
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Unconjugated
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Recombinant Human IL-6 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.