missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-6 Protein

A New rh IL-6 is Available! The new protein has ∼2 fold better activity and is HEK293 expressed!

Brand:  R&D Systems™ 206-IL-200/CF

837.99 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18748906

  • £1065.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.2-0.8 ng/mL.
Specifications

Specifications

Q75MH2
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
M.W. theroretical: 20.9 kDa
200 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Unconjugated
Reconstitute at 100-200 μg/mL in sterile PBS.
IL-6
Bioactivity
3569
IL-6
E. coli-derived human IL-6 protein Pro29-Met212
<0.10 EU / 1 μg of the protein by the LAL method.
IL6
Recombinant
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers