missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-6 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-34901-1mg
1828.00 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Assay #1:
- Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1 ng/mL, corresponding to a specific activity of >1.0 x 107 IU/mg.
Assay #2:
- Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using IL-6-dependent murine T1165 cells is less than 0.8 ng/mL, corresponding to a specific activity of >1.25 x 106 IU/mg.

Specifications
P05231, P05231, P05231 | |
PAGE, Bioactivity | |
3569 | |
1 mg | |
Less than 1.0 EU/ug of IL-6 as determined by LAL method. | |
IL6 | |
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
IL-6 |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. | |
20.8 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>95% by SDS-PAGE and HPLC |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human IL-6 Protein > 1 mg
Spot an opportunity for improvement?Share a Content Correction