missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL4 / Interleukin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_000580.1 |
---|---|
Gene ID (Entrez) | 3565 |
Name | IL4 / Interleukin-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085261
|
enQuireBio™
QP8557-ec-10ug |
10 μg | |||||||
16025271
|
enQuireBio™
QP8557-ec-50ug |
50 μg | |||||||
16075261
|
enQuireBio™
QP8557-ec-100ug |
100 μg | |||||||
16005271
|
enQuireBio™
QP8557-ec-200ug |
200 μg | |||||||
16015271
|
enQuireBio™
QP8557-ec-500ug |
500 μg | |||||||
16095261
|
enQuireBio™
QP8557-ec-1mg |
1 mg | |||||||
Specifications
NP_000580.1 | |
IL4 / Interleukin-4 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3565 | |
Research Use Only | |
IL4 | |
Recombinant Protein | |
E. coli | |
HKCDITLQEI IKTLNSLTEQ KTLCTELTVT DIFAASKNTT EKETFCRAAT VLRQFYSHHE KDTRCLGATA QQFHRHKQLI RFLKRLDRNL WGLAGLNSCP VKEANQSTLE NFLERLKTIM REKYSKCSS | |
Greater than 90% as determined by SDS-PAGE. |