missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCNAB2 Protein
A DNA sequence encoding the Homo sapiens (Human) KCNAB2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6252-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186789.1 | |
KCNAB2 Protein | |
Research Use Only | |
KCNAB2 | |
Recombinant Protein | |
E. coli | |
MYPESTTGSP ARLSLRQTGS PGMIYSTRYG SPKRQLQFYR NLGKSGLRVS CLGLGTWVTF GGQITDEMAE QLMTLAYDNG INLFDTAEVY AAGKAEVVLG NIIKKKGWRR SSLVITTKIF WGGKAETERG LSRKHIIEGL KASLERLQLE YVDVVFANRP DPNTPMEETV RAMTHVINQG MAMYWGTSRW SSMEIMEAYS VARQFNLTPP ICEQAEYHMF QREKVEVQLP ELFHKIGVGA MTWSPLACGI VSGKYDSGIP PYSRASLKGY QWLKDKILSE EGRRQQAKLK ELQAIAERLG CTLPQLAIAW CLRNEGVSSV LLGASNADQL MENIGAIQVL PKLSSSIIHE IDSILGNKPY SKKDYRS | |
Greater than 90% as determined by SDS-PAGE. |
8514 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |