missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KCNAB2 Protein

A DNA sequence encoding the Homo sapiens (Human) KCNAB2, was expressed in the hosts and tags indicated.

£145.46 - £1735.00

Specifications

Accession Number NP_001186789.1
Gene ID (Entrez) 8514
Name KCNAB2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16051930
View Documents Promotion Details
enQuireBio™
QP6252-EC-100UG
100 μg
£384.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16071930
View Documents Promotion Details
enQuireBio™
QP6252-EC-1MG
1 mg
£1735.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 05-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16061930
View Documents Promotion Details
enQuireBio™
QP6252-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16081930
View Documents Promotion Details
enQuireBio™
QP6252-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16091930
View Documents Promotion Details
enQuireBio™
QP6252-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16001940
View Documents Promotion Details
enQuireBio™
QP6252-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001186789.1
KCNAB2 Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
8514
Research Use Only
KCNAB2
Recombinant Protein
E. coli
MYPESTTGSP ARLSLRQTGS PGMIYSTRYG SPKRQLQFYR NLGKSGLRVS CLGLGTWVTF GGQITDEMAE QLMTLAYDNG INLFDTAEVY AAGKAEVVLG NIIKKKGWRR SSLVITTKIF WGGKAETERG LSRKHIIEGL KASLERLQLE YVDVVFANRP DPNTPMEETV RAMTHVINQG MAMYWGTSRW SSMEIMEAYS VARQFNLTPP ICEQAEYHMF QREKVEVQLP ELFHKIGVGA MTWSPLACGI VSGKYDSGIP PYSRASLKGY QWLKDKILSE EGRRQQAKLK ELQAIAERLG CTLPQLAIAW CLRNEGVSSV LLGASNADQL MENIGAIQVL PKLSSSIIHE IDSILGNKPY SKKDYRS
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications