missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LILRA5 Protein
A DNA sequence encoding the Homo sapiens (Human) LILRA5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP2841-ye-10ug
This item is not returnable.
View return policy
Specifications
NP_067073.1 | |
LILRA5 Protein | |
Research Use Only | |
LILRA5 | |
Recombinant Protein | |
Yeast | |
GNLSKATLWA EPGSVISRGN SVTIRCQGTL EAQEYRLVKE GSPEPWDTQN PLEPKNKARF SIPSMTEHHA GRYRCYYYSP AGWSEPSDPL ELVVTGFYNK PTLSALPSPV VTSGENVTLQ CGSRLRFDRF ILTEEGDHKL SWTLDSQLTP SGQFQALFPV GPVTPSHRWM LRCYGSRRHI LQVWSEPSDL LEIPVSGAAD NLSPSQNKSD SGTASHLQDY AVENLIR | |
Greater than 90% as determined by SDS-PAGE. |
353514 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |