missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PEBP1 Protein
A cDNA sequence encoding the PEBP1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | PEBP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962199
|
enQuireBio™
QP13001-10ug |
10 μg |
£99.15
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15982199
|
enQuireBio™
QP13001-50ug |
50 μg |
£155.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15972199
|
enQuireBio™
QP13001-1mg |
1 mg |
£1985.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PEBP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The PEBP1 protein solution contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPVDLSKWSG PLSLQEVDEQ PQHPLHVTYA GAAVDELGKV LTPTQVKNRP TSISWDGLDS GKLYTLVLTD PDAPSRKDPK YREWHHFLVV NMKGNDISSG TVLSDYVGSG PPKGTGLHRY VWLVYEQDRP LKCDEPILSN RSGDHRGKFK VASFRKKYEL RAPVAGTCYQ AEWDDYVPKL YEQLSGK | |
Greater than 95.0% as determined by SDS-PAGE. |