missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein BEX3 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein BEX3, was expressed in the hosts and tags indicated.
£145.46 - £1840.00
Specifications
Accession Number | NP_055195.1 |
---|---|
Gene ID (Entrez) | 27018 |
Name | Protein BEX3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083170
|
enQuireBio™
QP6419-ec-10ug |
10 μg | N/A | ||||||
16023180
|
enQuireBio™
QP6419-ec-50ug |
50 μg | N/A | ||||||
16073170
|
enQuireBio™
QP6419-ec-100ug |
100 μg |
£408.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16003180
|
enQuireBio™
QP6419-ec-200ug |
200 μg | N/A | ||||||
16013180
|
enQuireBio™
QP6419-ec-500ug |
500 μg | N/A | ||||||
16093170
|
enQuireBio™
QP6419-ec-1mg |
1 mg |
£1840.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_055195.1 | |
Protein BEX3 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
27018 | |
Research Use Only | |
NGFRAP1 | |
Recombinant Protein | |
E. coli | |
MANIHQENEE MEQPMQNGEE DRPLGGGEGH QPAGNRRGQA RRLAPNFRWA IPNRQINDGM GGDGDDMEIF MEEMREIRRK LRELQLRNCL RILMGELSNH HDHHDEFCLM P | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title