missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFBI (182 Amino Acids) Protein
A cDNA sequence encoding the TGFBI (182 Amino Acids) was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP14025-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
TGFBI (182 Amino Acids) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGTVMDVLKG DNRFSMLVAA IQSAGLTETL NREGVYTVFA PTNEAFRALP PRERSRLLGD AKELANILKY HIGDEILVSG GIGALVRLKS LQGDKLEVSL KNNVVSVNKE PVAEPDIMAT NGVVHVITNV LQPPANRPQE RGDELADSAL EIFKQASAFS RASQRSVRLA PVYQKLLERM KH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TGFBI 182 a. a. recombinant Human is formulated in 20mM Tris-HCl pH 8.0, 1mM EDTA, 0.1mM PMSF and 20% glycerol. |