missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFBI (182 Amino Acids) Protein
A cDNA sequence encoding the TGFBI (182 Amino Acids) was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | TGFBI (182 Amino Acids) Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995439
|
enQuireBio™
QP14025-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905449
|
enQuireBio™
QP14025-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915449
|
enQuireBio™
QP14025-EC-1MG |
1 mg |
£1985.00
1mg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TGFBI (182 Amino Acids) Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TGFBI 182 a. a. recombinant Human is formulated in 20mM Tris-HCl pH 8.0, 1mM EDTA, 0.1mM PMSF and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGTVMDVLKG DNRFSMLVAA IQSAGLTETL NREGVYTVFA PTNEAFRALP PRERSRLLGD AKELANILKY HIGDEILVSG GIGALVRLKS LQGDKLEVSL KNNVVSVNKE PVAEPDIMAT NGVVHVITNV LQPPANRPQE RGDELADSAL EIFKQASAFS RASQRSVRLA PVYQKLLERM KH | |
Greater than 95.0% as determined by SDS-PAGE. |