missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tryptophan 2,3-dioxygenase Protein
A DNA sequence encoding the Homo sapiens (Human) Tryptophan 2,3-dioxygenase, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_005642.1 |
---|---|
Gene ID (Entrez) | 6999 |
Name | Tryptophan 2,3-dioxygenase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085710
|
enQuireBio™
QP6770-ye-10ug |
10 μg | |||||||
16025720
|
enQuireBio™
QP6770-ye-50ug |
50 μg | |||||||
16075710
|
enQuireBio™
QP6770-ye-100ug |
100 μg |
£467.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16005720
|
enQuireBio™
QP6770-ye-200ug |
200 μg | |||||||
16015720
|
enQuireBio™
QP6770-ye-500ug |
500 μg |
£1265.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16095710
|
enQuireBio™
QP6770-ye-1mg |
1 mg | |||||||
Specifications
NP_005642.1 | |
Tryptophan 2,3-dioxygenase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6999 | |
Research Use Only | |
Tdo2 | |
Recombinant Protein | |
Yeast | |
MSGCPFLGNN FGYTFKKLPV EGSEEDKSQT GVNRASKGGL IYGNYLHLEK VLNAQELQSE TKGNKIHDEH LFIITHQAYE LWFKQILWEL DSVREIFQNG HVRDERNMLK VVSRMHRVSV ILKLLVQQFS ILETMTALDF NDFREYLSPA SGFQSLQFRL LENKIGVLQN MRVPYNRRHY RDNFKGEENE LLLKSEQEKT LLELVEAWLE RTPGLEPHGF NFWGKLEKNI TRGLEEEFIR IQAKEESEEK EEQVAEFQKQ KEVLLSLFDE KRHEHLLSKG ERRLSYRALQ GALMIYFYRE EPRFQVPFQL LTSLMDIDSL MTKWRYNHVC MVHRMLGSKA GTGGSSGYHY LRSTVSDRYK VFVDLFNLST YLIPRHWIPK MNPTIHKFLY TAEYCDSSYF SSDESD | |
Greater than 90% as determined by SDS-PAGE. |