missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2D4 Protein

A DNA sequence encoding the Homo sapiens (Human) UBE2D4, was expressed in the hosts and tags indicated.

£145.46 - £1735.00

Specifications

Accession Number NP_057067.1
Gene ID (Entrez) 51619
Name UBE2D4 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16003661
View Documents Promotion Details
enQuireBio™
QP8267-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16043661
View Documents Promotion Details
enQuireBio™
QP8267-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16093651
View Documents Promotion Details
enQuireBio™
QP8267-ec-100ug
100 μg
£384.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16023661
View Documents Promotion Details
enQuireBio™
QP8267-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16033661
View Documents Promotion Details
enQuireBio™
QP8267-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16013661
View Documents Promotion Details
enQuireBio™
QP8267-ec-1mg
1 mg
£1735.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_057067.1
UBE2D4 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
51619
Research Use Only
UBE2D4
Recombinant Protein
E. coli
MALKRIQKEL TDLQRDPPAQ CSAGPVGDDL FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TKIYHPNINS NGSICLDILR SQWSPALTVS KVLLSICSLL CDPNPDDPLV PEIAHTYKAD REKYNRLARE WTQKYAM
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications