missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UDP-glucuronosyltransferase 1-1 Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-glucuronosyltransferase 1-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9458-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000454.1 | |
UDP-glucuronosyltransferase 1-1 Protein | |
Research Use Only | |
UGT1A1 | |
Recombinant Protein | |
Yeast | |
HAGKILLIPV DGSHWLSMLG AIQQLQQRGH EIVVLAPDAS LYIRDGAFYT LKTYPVPFQR EDVKESFVSL GHNVFENDSF LQRVIKTYKK IKKDSAMLLS GCSHLLHNKE LMASLAESSF DVMLTDPFLP CSPIVAQYLS LPTVFFLHAL PCSLEFEATQ CPNPFSYVPR PLSSHSDHMT FLQRVKNMLI AFSQNFLCDV VYSPYATLAS EFLQREVTVQ DLLSSASVWL FRSDFVKDYP RPIMPNMVFV GGINCLHQNP LSQEFEAYIN ASGEHGIVVF SLGSMVSEIP EKKAMAIADA LGKIPQTVLW RYTGTRPSNL ANNTILVKWL PQNDLLGHPM TRAFITHAGS HGVYESICNG VPMVMMPLFG DQMDNAKRME TKGAGVTLNV LEMTSEDLEN ALKAVINDKS YKENIMRLSS LHKDRPVEPL DLAVFWVEFV MRHKGAPHLR PAAHDLTWYQ YHSLDVIGFL LAVVLTVAFI TFKCCAYGYR KCLGKKGRVK KAHKSKTH | |
Greater than 90% as determined by SDS-PAGE. |
54658 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |