missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UDP-glucuronosyltransferase 1-1 Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-glucuronosyltransferase 1-1, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_000454.1 |
---|---|
Gene ID (Entrez) | 54658 |
Name | UDP-glucuronosyltransferase 1-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038341
|
enQuireBio™
QP9458-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078341
|
enQuireBio™
QP9458-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048341
|
enQuireBio™
QP9458-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068341
|
enQuireBio™
QP9458-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088341
|
enQuireBio™
QP9458-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058341
|
enQuireBio™
QP9458-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000454.1 | |
UDP-glucuronosyltransferase 1-1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
54658 | |
Research Use Only | |
UGT1A1 | |
Recombinant Protein | |
Yeast | |
HAGKILLIPV DGSHWLSMLG AIQQLQQRGH EIVVLAPDAS LYIRDGAFYT LKTYPVPFQR EDVKESFVSL GHNVFENDSF LQRVIKTYKK IKKDSAMLLS GCSHLLHNKE LMASLAESSF DVMLTDPFLP CSPIVAQYLS LPTVFFLHAL PCSLEFEATQ CPNPFSYVPR PLSSHSDHMT FLQRVKNMLI AFSQNFLCDV VYSPYATLAS EFLQREVTVQ DLLSSASVWL FRSDFVKDYP RPIMPNMVFV GGINCLHQNP LSQEFEAYIN ASGEHGIVVF SLGSMVSEIP EKKAMAIADA LGKIPQTVLW RYTGTRPSNL ANNTILVKWL PQNDLLGHPM TRAFITHAGS HGVYESICNG VPMVMMPLFG DQMDNAKRME TKGAGVTLNV LEMTSEDLEN ALKAVINDKS YKENIMRLSS LHKDRPVEPL DLAVFWVEFV MRHKGAPHLR PAAHDLTWYQ YHSLDVIGFL LAVVLTVAFI TFKCCAYGYR KCLGKKGRVK KAHKSKTH | |
Greater than 90% as determined by SDS-PAGE. |