missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Adck2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Adck2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7772-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_849204.1 | |
Adck2 Protein | |
Research Use Only | |
Adck2 | |
Recombinant Protein | |
E. coli | |
FETREPVGSG CVAQVYKAFA SISLLEEDRI WRLGELSAPG TRAVVMQREP FMKDRKPSEN LADEAFLEKL LLPKADLGGS EVGVSQAPWH LPKSDHLIPV AVKVLHPGLL SQVSMDLLLM KIGSKALGLL PGVKWLSLPE IVEEFEKLMV QQTDLRYEAQ NLEHFQHNFQ DMASVKFPTP LRPLITRDIL VETYEESVPV SSYQQAGIPT DLKRKIAQLG INMLLKMIFV DNFVHGDLHP GNILVQGADG VSPSLEMQQQ QVNVCDTLVA TIAPALCPLR LVLLDAGIVA KLQASDLRNF RAVFQAVVMG QGHRVAELML HHAQANECRD VERFKAEMAT LVTQARKNIV TLEKLHVSSL LSSVFKLLMT HKVKLESNFA SIVVAIMVLE GLGRSLDPTL DILEAAKPFL | |
Greater than 90% as determined by SDS-PAGE. |
57869 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |