missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Adck2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Adck2, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_849204.1 |
---|---|
Gene ID (Entrez) | 57869 |
Name | Adck2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16060691
|
enQuireBio™
QP7772-ec-10ug |
10 μg | N/A | ||||||
16000701
|
enQuireBio™
QP7772-ec-50ug |
50 μg | N/A | ||||||
16050691
|
enQuireBio™
QP7772-ec-100ug |
100 μg |
£560.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16080691
|
enQuireBio™
QP7772-ec-200ug |
200 μg | N/A | ||||||
16090691
|
enQuireBio™
QP7772-ec-500ug |
500 μg | N/A | ||||||
16070691
|
enQuireBio™
QP7772-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_849204.1 | |
Adck2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
57869 | |
Research Use Only | |
Adck2 | |
Recombinant Protein | |
E. coli | |
FETREPVGSG CVAQVYKAFA SISLLEEDRI WRLGELSAPG TRAVVMQREP FMKDRKPSEN LADEAFLEKL LLPKADLGGS EVGVSQAPWH LPKSDHLIPV AVKVLHPGLL SQVSMDLLLM KIGSKALGLL PGVKWLSLPE IVEEFEKLMV QQTDLRYEAQ NLEHFQHNFQ DMASVKFPTP LRPLITRDIL VETYEESVPV SSYQQAGIPT DLKRKIAQLG INMLLKMIFV DNFVHGDLHP GNILVQGADG VSPSLEMQQQ QVNVCDTLVA TIAPALCPLR LVLLDAGIVA KLQASDLRNF RAVFQAVVMG QGHRVAELML HHAQANECRD VERFKAEMAT LVTQARKNIV TLEKLHVSSL LSSVFKLLMT HKVKLESNFA SIVVAIMVLE GLGRSLDPTL DILEAAKPFL | |
Greater than 90% as determined by SDS-PAGE. |