missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse C1q-related factor Protein

A DNA sequence encoding the Mus musculus (Mouse) C1q-related factor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9018-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017461

  • £560.00 / 100µg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Specifications

Specifications

C1q-related factor Protein
Research Use Only
C1ql1
Recombinant Protein
Yeast
GPDGHYEMLG TCRMVCDPYP ARGPGAGARS DGGDALSEQS GAPPPSTLVQ GPQGKPGRTG KPGPPGPPGD RGPPGPVGPP GEKGEPGKPG PPGLPGSGGS GAISTATYTT VPRVAFYAGL KNPHEGYEVL KFDDVVTNLG NNYDAASGKF TCNIPGTYFF TYHVLMRGGD GTSMWADLCK NGQVRASAIA QDADQNYDYA SNSVILHLDA GDEVFIKLDG GKAHGGNSNK YSTFSGFIIY SD
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers