missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C1q-related factor Protein
A DNA sequence encoding the Mus musculus (Mouse) C1q-related factor, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Name | C1q-related factor Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | C1ql1 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017461
|
enQuireBio™
QP9018-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057461
|
enQuireBio™
QP9018-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027461
|
enQuireBio™
QP9018-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047461
|
enQuireBio™
QP9018-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067461
|
enQuireBio™
QP9018-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037461
|
enQuireBio™
QP9018-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
C1q-related factor Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
C1ql1 | |
Recombinant Protein | |
Yeast | |
GPDGHYEMLG TCRMVCDPYP ARGPGAGARS DGGDALSEQS GAPPPSTLVQ GPQGKPGRTG KPGPPGPPGD RGPPGPVGPP GEKGEPGKPG PPGLPGSGGS GAISTATYTT VPRVAFYAGL KNPHEGYEVL KFDDVVTNLG NNYDAASGKF TCNIPGTYFF TYHVLMRGGD GTSMWADLCK NGQVRASAIA QDADQNYDYA SNSVILHLDA GDEVFIKLDG GKAHGGNSNK YSTFSGFIIY SD | |
Greater than 90% as determined by SDS-PAGE. |