missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C1q-related factor Protein
A DNA sequence encoding the Mus musculus (Mouse) C1q-related factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9018-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
C1q-related factor Protein | |
Research Use Only | |
C1ql1 | |
Recombinant Protein | |
Yeast | |
GPDGHYEMLG TCRMVCDPYP ARGPGAGARS DGGDALSEQS GAPPPSTLVQ GPQGKPGRTG KPGPPGPPGD RGPPGPVGPP GEKGEPGKPG PPGLPGSGGS GAISTATYTT VPRVAFYAGL KNPHEGYEVL KFDDVVTNLG NNYDAASGKF TCNIPGTYFF TYHVLMRGGD GTSMWADLCK NGQVRASAIA QDADQNYDYA SNSVILHLDA GDEVFIKLDG GKAHGGNSNK YSTFSGFIIY SD | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |