missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse Cardiotrophin-1/CT-1 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-35170-10ug
Additional Details : Weight : 0.00970kg
Description
Cardiotrophin-1/CT-1 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF1 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 10^6 IU/mg.Specifications
Q60753, Q60753, Q60753 | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, 300 mM NaCl, pH 8.5. | |
M.W. (Theoretical): 21.5 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1 | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C | |
>95%, by SDS-PAGE and HPLC |
PAGE, Bioactivity | |
1489 | |
10 μg | |
Less than 1 EU/μg of Cardiotrophin-1/CT-1 as determined by LAL method. | |
CTF1 | |
Biologically Active Proteins | |
Cardiotrophin-1/CT-1 |