missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse Cardiotrophin-1/CT-1 Protein
Recombinant Protein
£171.70 - £3727.76
Specifications
Accession Number | Q60753, Q60753, Q60753 |
---|---|
For Use With (Application) | PAGE, Bioactivity |
Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, 300 mM NaCl, pH 8.5. |
Gene ID (Entrez) | 1489 |
Molecular Weight (g/mol) | M.W. (Theoretical): 21.5 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18404152
|
Novus Biologicals™
NBP2-35170-500UG |
500 μg |
£2133.85
500µg |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18448051
|
Novus Biologicals™
NBP2-35170-100UG |
100 μg |
£966.50
100µg |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18437401
|
Novus Biologicals™
NBP2-35170-10UG |
10 μg |
£171.70
10µg |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18476491
|
Novus Biologicals™
NBP2-35170-1MG |
1mg |
£3727.76
1mg |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cardiotrophin-1/CT-1 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF1 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 10^6 IU/mg.Specifications
Q60753, Q60753, Q60753 | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, 300 mM NaCl, pH 8.5. | |
M.W. (Theoretical): 21.5 kDa | |
Less than 1 EU/μg of Cardiotrophin-1/CT-1 as determined by LAL method. | |
CTF1 | |
Biologically Active Proteins | |
Cardiotrophin-1/CT-1 |
PAGE, Bioactivity | |
1489 | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1 | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C | |
>95%, by SDS-PAGE and HPLC |