missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 3210-C-025/CF
261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) is derived from E. coli. The Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
8.3 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
20305 | |
CCL6/C10 Truncated, aa 42-116 | |
E. coli-derived mouse CCL6/C10 protein Gly42-Ala116 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |