missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£333.00
Specifications
Gene ID (Entrez) | 20305 |
---|---|
Molecular Weight (g/mol) | 8.3 kDa |
Name | CCL6/C10 Truncated, aa 42-116 |
Quantity | 25 μg |
Source | E. coli-derived mouse CCL6/C10 protein Gly42-Ala116 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18176400
|
R&D Systems™
3210-C-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£333.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18305557
|
R&D Systems™
3210-C-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£333.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein is derived from E. coli. The Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein has been validated for the following applications: Bioactivity.Specifications
20305 | |
CCL6/C10 Truncated, aa 42-116 | |
E. coli-derived mouse CCL6/C10 protein Gly42-Ala116 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
8.3 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |