missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MUP1 Protein
A cDNA sequence encoding the MUP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12754-1mg
This item is not returnable.
View return policy
Specifications
MUP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEEASSTG RNFNVEKING EWHTIILASD KREKIEDNGN FRLFLEQIHV LENSLVLKFH TVRDEECSEL SMVADKTEKA GEYSVTYDGF NTFTIPKTDY DNFLMAHLIN EKDGETFQLM GLYGREPDLS SDIKERFAQL CEKHGILREN IIDLSNANRC LQARE | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
The MUP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |