missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MUP1 Protein
A cDNA sequence encoding the MUP1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | MUP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951439
|
enQuireBio™
QP12754-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941439
|
enQuireBio™
QP12754-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15931439
|
enQuireBio™
QP12754-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
MUP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
The MUP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEEASSTG RNFNVEKING EWHTIILASD KREKIEDNGN FRLFLEQIHV LENSLVLKFH TVRDEECSEL SMVADKTEKA GEYSVTYDGF NTFTIPKTDY DNFLMAHLIN EKDGETFQLM GLYGREPDLS SDIKERFAQL CEKHGILREN IIDLSNANRC LQARE | |
Greater than 95% as determined by SDS-PAGE. |