missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Phosphoserine aminotransferase Protein
A DNA sequence encoding the Mus musculus (Mouse) Phosphoserine aminotransferase, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_803155.1 |
---|---|
Gene ID (Entrez) | 107272 |
Name | Phosphoserine aminotransferase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082311
|
enQuireBio™
QP8039-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16002321
|
enQuireBio™
QP8039-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092311
|
enQuireBio™
QP8039-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012321
|
enQuireBio™
QP8039-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022321
|
enQuireBio™
QP8039-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032321
|
enQuireBio™
QP8039-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_803155.1 | |
Phosphoserine aminotransferase Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
107272 | |
Research Use Only | |
Psat1 | |
Recombinant Protein | |
E. coli | |
MEATKQVVNF GPGPAKLPHS VLLEIQKQLL DYRGLGISVL EMSHRSSDFA KIIGNTENLV RELLAVPNNY KVIFVQGGGS GQFSAVPLNL IGLKAGRSAD YVVTGAWSAK AAEEAKKFGT VNIVHPKLGS YTKIPDPSTW NLNPDASYVY FCANETVHGV EFDFVPDVKG AVLVCDMSSN FLSRPVDVSK FGVIFAGAQK NVGSAGVTVV IVRDDLLGFS LRECPSVLDY KVQAGNNSLY NTPPCFSIYV MGMVLEWIKN NGGAAAMEKL SSIKSQMIYE IIDNSQGFYV CPVERQNRSR MNIPFRIGNA KGDEALEKRF LDKAVELNMI SLKGHRSVGG IRASLYNAVT TEDVEKLAAF MKNFLEMHQL | |
Greater than 90% as determined by SDS-PAGE. |