Learn More
R&D Systems™ Recombinant Mouse sTNF RII/TNFRSF1B Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 426-R2-050
This item is not returnable.
View return policy
Description
The Recombinant Mouse sTNF RII/TNFRSF1B Protein is derived from E. coli. The Recombinant Mouse sTNF RII/TNFRSF1B Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
25 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
21938 | |
TNF RII/TNFRSF1B | |
E. coli-derived mouse TNF RII/TNFRSF1B protein Val23-Gly258 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |