missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse sTNF RII/TNFRSF1B Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£416.84
Specifications
Gene ID (Entrez) | 21938 |
---|---|
Molecular Weight (g/mol) | 25 kDa |
Name | TNF RII/TNFRSF1B |
Quantity | 50μg |
Source | E. coli-derived mouse TNF RII/TNFRSF1B protein Val23-Gly258 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18330368
|
R&D Systems™
426-R2-050 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£416.84
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18347227
|
R&D Systems™
426-R2-050/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£416.84
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse sTNF RII/TNFRSF1B Protein is derived from E. coli. The Recombinant Mouse sTNF RII/TNFRSF1B Protein has been validated for the following applications: Bioactivity.Specifications
21938 | |
TNF RII/TNFRSF1B | |
E. coli-derived mouse TNF RII/TNFRSF1B protein Val23-Gly258 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
25 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |