Learn More
R&D Systems™ Recombinant Mouse sTNF RII/TNFRSF1B Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 426-R2-050/CF
 View more versions of this product
Product Code. 18347227
  • £416.84 / 50µg
Explore more special offers
Description
Description
The Recombinant Mouse sTNF RII/TNFRSF1B Protein is derived from E. coli. The Recombinant Mouse sTNF RII/TNFRSF1B Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
25 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
21938
TNF RII/TNFRSF1B
E. coli-derived mouse TNF RII/TNFRSF1B protein Val23-Gly258
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.