missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tryptophan Hydroxylase 1 / TPH1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Tryptophan Hydroxylase 1 / TPH1, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001129556.1 |
---|---|
Gene ID (Entrez) | 21990 |
Name | Tryptophan Hydroxylase 1 / TPH1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16076100
|
enQuireBio™
QP6826-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096100
|
enQuireBio™
QP6826-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086100
|
enQuireBio™
QP6826-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006110
|
enQuireBio™
QP6826-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016110
|
enQuireBio™
QP6826-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026110
|
enQuireBio™
QP6826-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001129556.1 | |
Tryptophan Hydroxylase 1 / TPH1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
21990 | |
Research Use Only | |
Tph1 | |
Recombinant Protein | |
E. coli | |
MIEDNKENKE NKDHSSERGR VTLIFSLENE VGGLIKVLKI FQENHVSLLH IESRKSKQRN SEFEIFVDCD ISREQLNDIF PLLKSHATVL SVDSPDQLTA KEDVMETVPW FPKKISDLDF CANRVLLYGS ELDADHPGFK DNVYRRRRKY FAELAMNYKH GDPIPKIEFT EEEIKTWGTI FRELNKLYPT HACREYLRNL PLLSKYCGYR EDNIPQLEDV SNFLKERTGF SIRPVAGYLS PRDFLSGLAF RVFHCTQYVR HSSDPLYTPE PDTCHELLGH VPLLAEPSFA QFSQEIGLAS LGASEETVQK LATCYFFTVE FGLCKQDGQL RVFGAGLLSS ISELKHALSG HAKVKPFDPK IACKQECLIT SFQDVYFVSE SFEDAKEKMR EFAKTVKRPF GLKYNPYTQS VQVLRDTKSI TSAMNELRYD LDVISDALAR VTRWPSV | |
Greater than 90% as determined by SDS-PAGE. |