missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Carbonic Anhydrase II Protein
A cDNA sequence encoding the Carbonic Anhydrase II was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11275-25ug
Additional Details : Weight : 0.01000kg
Specifications
944832 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKDIDTLISN NALWSKMLVE EDPGFFEKLAQAQKPRFLWI GCSDSRVPAE RLTGLEPGEL FVHRNVANLV IHTDLNCLSV VQYAVDVLEV EHIIICGHYG CGGVQAAVEN PELGLINNWL HIRDIWFKH SSLLGEMPQE RRLDTLCELN VMEQVYNLGH STIMQSAWKR GQKVTIHGWA YGIHDGLLRD LDVTATNRET LEQRYRHGIS NLKLKHANHK | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Carbonic Anhydrase II Protein | |
Research Use Only | |
can | |
Other | |
His | |
The Carbonic Anhydrase 2 enzyme is supplied in 20mM Tris pH 8 and 1mM DTT. |