missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Carbonic Anhydrase II Protein
A cDNA sequence encoding the Carbonic Anhydrase II was constructed and used to recombinantly synthesize the protein.
£99.15 - £3085.00
Specifications
Gene ID (Entrez) | 944832 |
---|---|
Name | Carbonic Anhydrase II Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | can |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976688
|
enQuireBio™
QP11275-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966688
|
enQuireBio™
QP11275-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956688
|
enQuireBio™
QP11275-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
944832 | |
Research Use Only | |
can | |
Other | |
His | |
The Carbonic Anhydrase 2 enzyme is supplied in 20mM Tris pH 8 and 1mM DTT. |
Carbonic Anhydrase II Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKDIDTLISN NALWSKMLVE EDPGFFEKLAQAQKPRFLWI GCSDSRVPAE RLTGLEPGEL FVHRNVANLV IHTDLNCLSV VQYAVDVLEV EHIIICGHYG CGGVQAAVEN PELGLINNWL HIRDIWFKH SSLLGEMPQE RRLDTLCELN VMEQVYNLGH STIMQSAWKR GQKVTIHGWA YGIHDGLLRD LDVTATNRET LEQRYRHGIS NLKLKHANHK | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |