missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Recombinant Protein A-Sepharose™ 4B
Recombinant Protein A-Sepharose™ 4B
£324.00 - £920.00
Specifications
Product Type | Protein A Sepharose |
---|---|
Column Type | Affinity |
Stationary Phase | Protein A |
Form | Suspension |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10252783
|
Novex™
101141 |
5 mL |
£324.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10716713
|
Novex™
101142 |
25 mL |
£920.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This product is prepared by covalently coupling highly purified Staph. aureus Cowan strain Protein A produced in E. Coli to Sepharose™ 4B.This product is supplied as a 50% suspension consisting of recombinant Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% ).
Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
Specifications
Protein A Sepharose | |
Affinity | |
Protein A | |
Suspension |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novex™ Recombinant Protein A-Sepharose™ 4B