missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein A-Sepharose™ 4B

Recombinant Protein A-Sepharose™ 4B

Brand:  Novex™ 101142

 View more versions of this product

Product Code. 10716713

  • £920.00 / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

This product is prepared by covalently coupling highly purified Staph. aureus Cowan strain Protein A produced in E. Coli to Sepharose™ 4B.

This product is supplied as a 50% suspension consisting of recombinant Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% ).

Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein A Sepharose
Protein A
25 mL
Affinity
2°C to 8°C. Do Not Freeze.
This product is supplied as a 50% suspension (25 ml Recombinant Protein A-Sepharose™ 4B + 25 ml PBS) Wet Ice
Suspension
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAC411525 Certificate TypeCertificate of Analysis Date15/04/2025 Catalog Number
Lot NumberYB360085 Certificate TypeCertificate of Analysis Date08/07/2023 Catalog Number101142
Lot NumberXG335957 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number101142
Lot NumberXB332388 Certificate TypeCertificate of Analysis Date17/05/2022 Catalog Number101142
Lot NumberWH325163 Certificate TypeCertificate of Analysis Date09/10/2021 Catalog Number101142
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novex™ Recombinant Protein A-Sepharose™ 4B > 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.