missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Universal Nuclease for Cell Lysis

Ideal for a wide variety of applications where complete digestion of nucleic acids is needed and can be used in conjunction with cell lysis buffers.

Brand:  Thermo Scientific™ 88700

77.43 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 12146490

  • £83.50 / 5ku

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Universal Nuclease for Cell Lysis is ideal for a wide variety of applications where complete digestion of nucleic acids is needed and can be used in conjunction with cell lysis buffers.

Pierce Universal Nuclease for Cell Lysis is a genetically engineered endonuclease from Serratia marcescens. The enzyme is produced and purified from E. coli and consists of two identical 30-kDa subunits with two critical disulfide bonds. This indiscriminate endonuclease degrades single-stranded, double-stranded, linear and circular DNA and RNA and is effective over a wide range of temperatures and pH. This enzyme has very high specific activity (100-fold greater than DNase I) and has increased thermal stability compared to other nucleases. Pierce Universal Nuclease is ≥99% pure enzyme, is free of any measurable protease activity and is supplied at a concentration of 250 U/μL. Pierce Universal Nuclease for Cell Lysis is identical in performance to Benzonase™ Nuclease by EMD Merck™.

Highlights:

  • Broad spectrum – degrades all forms of DNA and RNA
  • Highest-quality enzyme – nuclease is at least 99% pure, as tested by SDS-PAGE
  • Robust activity – 100-fold greater specific activity than DNase I
  • Versatile – can be used with a wide variety of cell lysis reagents

Recommended for:

Use with B-PER, Y-PER or other commercial or homebrew cell lysis reagents and/or mechanical disruption to reduce viscosity in protein extracts; Remove DNA and RNA from recombinant protein preparations prior to downstream processing

Storage Requirements

Store in a cool, dry, well-ventilated area, protected from direct sunlight

TRUSTED_SUSTAINABILITY
Specifications

Specifications

At least 99% pure enzyme at 250 U/uL in 50% glycerol (v/v), 20 mM NaCl, 2 mM MgCl2, 20 mM Tris, pH 8.0
Cell Lysis Enzyme
Liquid
Enzyme for Cell Lysis
200 mL Lysate
Pierce Universal Nuclease for Cell Lysis
Store in a cool, dry, well-ventilated area, protected from direct sunlight.
Pierce™
Nuclease
5 kU
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2865324 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number88700
Lot Number3014249 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number88700
Lot Number2941602 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number88700
Lot Number3103334 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number88700
Lot Number2941724 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number88700
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Universal Nuclease for Cell Lysis > 5 kU

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.