Learn More
Thermo Scientific™ Lysozyme
Degrade unwanted bacterial cell wall in protein or nucleic acid extracts
Brand: Thermo Scientific™ 89833
131.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Lysozyme is an enzyme used to break down bacterial cell walls to improve protein or nucleic acid extraction efficiency. Lysozymes (muramidases) are a family of enzymes with antimicrobial activity characterized by the ability to damage the cell wall of bacteria. The enzyme acts by catalyzing the hydrolysis of 1,4-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycans and between the N-acetyl-D-glucosamine residues in chitodextrins. Although hen egg white lysozyme is most effective for the lysis of gram-positive bacteria, it also facilitates the lysis of gram-negative bacteria such as Salmonella and Shigella. The lysis of E. coli is especially improved by the addition of both lysozyme and a nucleases such as DNase I.
Highlights:
- Bacterial cell wall lytic enzyme that improves inclusion body protein purification
- Compatible with Thermo Scientific Pierce Cell Lysis Reagents
- Available in two convenient formats: lyophilized powder for large volume applications (Part No. 89833) and convenient glycerol freezer stock (Part No. 90082)
Properties of lysozyme:
- Enzymatic function: Catalyzes the hydrolysis of 1,4-β-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycans and between N-acetyl-D-glucosamine residues in chitodextrins
- Molecular weight: 14.388 kDa
- Extinction coefficient: ∽26.4 at 280nm
- Isoelectric point: pH 11.0
- Unit definition: 1 unit is that amount of enzyme needed to catalyze a decrease in absorbance at 450nm of 0.001/min at 25 degrees at pH 6.24 in a 1cm cuvette due to lysis using a ∼0.25mg/mL suspension of Micrococcus luteus.

Specifications
Cell Lysis Enzyme | |
Lyophilized | |
Lysozyme (Muramidase) |
Upon receipt, store at -20°C in a nonfrost-free freezer. Product shipped with ice. | |
Enzyme for Cell Lysis | |
5 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.