Learn More
Thermo Scientific™ Pierce™ Universal Nuclease for Cell Lysis
Ideal for a wide variety of applications where complete digestion of nucleic acids is needed and can be used in conjunction with cell lysis buffers.
£83.50 - £546.00
Specifications
Product Type | Cell Lysis Enzyme |
---|---|
Content And Storage | Store in a cool, dry, well-ventilated area, protected from direct sunlight. |
Form | Liquid |
Product Line | Pierce™ |
Reagent Type | Enzyme for Cell Lysis |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12146490
|
Thermo Scientific™
88700 |
5 kU |
£83.50
5ku |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12391963
|
Thermo Scientific™
88702 |
100 kU |
£546.00
100ku |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12156490
|
Thermo Scientific™
88701 |
25 kU |
£250.00
25ku |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Pierce Universal Nuclease for Cell Lysis is ideal for a wide variety of applications where complete digestion of nucleic acids is needed and can be used in conjunction with cell lysis buffers.
Pierce Universal Nuclease for Cell Lysis is a genetically engineered endonuclease from Serratia marcescens. The enzyme is produced and purified from E. coli and consists of two identical 30-kDa subunits with two critical disulfide bonds. This indiscriminate endonuclease degrades single-stranded, double-stranded, linear and circular DNA and RNA and is effective over a wide range of temperatures and pH. This enzyme has very high specific activity (100-fold greater than DNase I) and has increased thermal stability compared to other nucleases. Pierce Universal Nuclease is ≥99% pure enzyme, is free of any measurable protease activity and is supplied at a concentration of 250 U/μL. Pierce Universal Nuclease for Cell Lysis is identical in performance to Benzonase™ Nuclease by EMD Merck™.
Highlights:
- Broad spectrum – degrades all forms of DNA and RNA
- Highest-quality enzyme – nuclease is at least 99% pure, as tested by SDS-PAGE
- Robust activity – 100-fold greater specific activity than DNase I
- Versatile – can be used with a wide variety of cell lysis reagents
Recommended for:
Use with B-PER, Y-PER or other commercial or homebrew cell lysis reagents and/or mechanical disruption to reduce viscosity in protein extracts; Remove DNA and RNA from recombinant protein preparations prior to downstream processing
Storage Requirements
Store in a cool, dry, well-ventilated area, protected from direct sunlight
Specifications
Cell Lysis Enzyme | |
Liquid | |
Enzyme for Cell Lysis |
Store in a cool, dry, well-ventilated area, protected from direct sunlight. | |
Pierce™ | |
Nuclease |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.