missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Carbonic anhydrase 1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Carbonic anhydrase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5741-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957429

  • £2480.00 / 1mg
Estimated Shipment: 02-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001101130.1
Carbonic anhydrase 1 Protein
Research Use Only
Ca1
Recombinant Protein
Yeast
ASADWGYDSK NGPDQWSKLY PIANGNNQSP IDIKTSEAKH DSSLKPVSVS YNPATAKEIV NVGHSFHVVF DDSSNQSVLK GGPLADSYRL TQFHFHWGNS NDHGSEHTVD GAKYSGELHL VHWNSAKYSS AAEAISKADG LAIIGVLMKV GPANPNLQKV LDALSSVKTK GKRAPFTNFD PSSLLPSSLD YWTYFGSLTH PPLHESVTWV ICKESISLSP EQLAQLRGLL SSAEGEPAVP VLSNHRPPQP LKGRTVRASF
Greater than 90% as determined by SDS-PAGE.
310218
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers