missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Carbonic anhydrase 1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Carbonic anhydrase 1, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001101130.1 |
---|---|
Gene ID (Entrez) | 310218 |
Name | Carbonic anhydrase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15937429
|
enQuireBio™
QP5741-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957429
|
enQuireBio™
QP5741-YE-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947429
|
enQuireBio™
QP5741-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15967429
|
enQuireBio™
QP5741-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15977429
|
enQuireBio™
QP5741-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15987429
|
enQuireBio™
QP5741-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001101130.1 | |
Carbonic anhydrase 1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
310218 | |
Research Use Only | |
Ca1 | |
Recombinant Protein | |
Yeast | |
ASADWGYDSK NGPDQWSKLY PIANGNNQSP IDIKTSEAKH DSSLKPVSVS YNPATAKEIV NVGHSFHVVF DDSSNQSVLK GGPLADSYRL TQFHFHWGNS NDHGSEHTVD GAKYSGELHL VHWNSAKYSS AAEAISKADG LAIIGVLMKV GPANPNLQKV LDALSSVKTK GKRAPFTNFD PSSLLPSSLD YWTYFGSLTH PPLHESVTWV ICKESISLSP EQLAQLRGLL SSAEGEPAVP VLSNHRPPQP LKGRTVRASF | |
Greater than 90% as determined by SDS-PAGE. |