missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat Pentraxin 2/SAP Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 1895-SA-050/CF
This item is not returnable.
View return policy
Description
The Recombinant Rat Pentraxin 2/SAP Protein is derived from NS0. The Recombinant Rat Pentraxin 2/SAP Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 . | |
25 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
29339 | |
Pentraxin 2/SAP | |
Mouse myeloma cell line,NS0-derived rat Pentraxin 2/SAP protein Gln21-Ser228 with a C-terminal 10-His tag | |
Noncovalently-linked homopentamer | |
Recombinant |