missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat Pentraxin 2/SAP Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£344.48
Specifications
Gene ID (Entrez) | 29339 |
---|---|
Molecular Weight (g/mol) | 25 kDa |
Name | Pentraxin 2/SAP |
Quantity | 50μg |
Source | Mouse myeloma cell line,NS0-derived rat Pentraxin 2/SAP protein Gln21-Ser228 with a C-terminal 10-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18107370
|
R&D Systems™
1895-SA-050 |
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein. |
£344.48
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18389647
|
R&D Systems™
1895-SA-050/CF |
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 . |
£344.48
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Rat Pentraxin 2/SAP Protein is derived from NS0. The Recombinant Rat Pentraxin 2/SAP Protein has been validated for the following applications: Binding Activity.Specifications
29339 | |
Pentraxin 2/SAP | |
Mouse myeloma cell line,NS0-derived rat Pentraxin 2/SAP protein Gln21-Ser228 with a C-terminal 10-His tag | |
Noncovalently-linked homopentamer | |
Recombinant |
25 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |