missing translation for 'onlineSavingsMsg'
Learn More

Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 568, Clone: CTK0101 and CTK0102, Proteintech ChromoTek

Alpaca Monoclonal Antibody

Brand:  Chromotek SRBAF568-1-100

Additional Details : Weight : 0.00240kg

 View more versions of this product

Product Code. 17383493

  • £277.00 / 100µL
Estimated Shipment: 23-07-2024
to see stock.

Description

Description

Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.

Specifications

Specifications

Recombinant VHH
Monoclonal
Alexa Fluor 568
100 μL
Human, Rabbit
Immunofluorescence, Western Blot, Flow Cytometry
CTK0101 and CTK0102
Alpaca
Secondary
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions