missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 568, Clone: CTK0101 and CTK0102, Proteintech ChromoTek
Alpaca Monoclonal Antibody
£76.00 - £277.00
Specifications
Antigen | Recombinant VHH |
---|---|
Clone | CTK0101 and CTK0102 |
Applications | Immunofluorescence, Western Blot, Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 568 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17373493
|
Chromotek
SRBAF568-1-10 |
10 μL |
£76.00
10µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17383493
|
Chromotek
SRBAF568-1-100 |
100 μL |
£277.00
100µL |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Specifications
Recombinant VHH | |
Immunofluorescence, Western Blot, Flow Cytometry | |
Alexa Fluor 568 | |
Human, Rabbit | |
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
CTK0101 and CTK0102 | |
Monoclonal | |
Alpaca | |
Secondary |