missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 647, Clone: CTK0101 and CTK0102, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand: Chromotek srbAF647-1-100
This item is not returnable.
View return policy
Description
Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Specifications
Recombinant VHH | |
Monoclonal | |
Alexa Fluor 647 | |
100 μL | |
Human, Rabbit |
Immunofluorescence, Western Blot, Flow Cytometry | |
CTK0101 and CTK0102 | |
Alpaca | |
Secondary | |
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 647, Clone: CTK0101 and CTK0102, Proteintech ChromoTek > 100 μL
Spot an opportunity for improvement?Share a Content Correction