missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 647, Clone: CTK0101 and CTK0102, Proteintech ChromoTek
Alpaca Monoclonal Antibody
£76.00 - £276.00
Specifications
Antigen | Recombinant VHH |
---|---|
Clone | CTK0101 and CTK0102 |
Applications | Immunofluorescence, Western Blot, Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17393493
|
Chromotek
srbAF647-1-10 |
10 μL |
£76.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17303503
|
Chromotek
srbAF647-1-100 |
100 μL |
£276.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Specifications
Recombinant VHH | |
Immunofluorescence, Western Blot, Flow Cytometry | |
Alexa Fluor 647 | |
Human, Rabbit | |
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
CTK0101 and CTK0102 | |
Monoclonal | |
Alpaca | |
Secondary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 647, Clone: CTK0101 and CTK0102, Proteintech ChromoTek